Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Trypsin 3/PRSS3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3565SP
This item is not returnable.
View return policy
Description
Trypsin 3/PRSS3 Polyclonal specifically detects Trypsin 3/PRSS3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Trypsin 3/PRSS3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsin, Mesotrypsinogen, MTG, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS3, PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin 4, Trypsin III, Trypsin IV, trypsin-3, Trypsinogen 15, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
Goat | |
Affinity Purified | |
RUO | |
5646 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
NP_035775 | |
PRSS3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Trypsin 3 Phe16-Asn246 Accession # NP_035775 | |
25 μg | |
Primary | |
Detects mouse Trypsin 3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Trypsin 1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction