Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Trypsin 3/PRSS3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB35651
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB35651 100 μg
MAB35651SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB35651 Supplier R&D Systems Supplier No. MAB35651
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Trypsin 3/PRSS3 Monoclonal specifically detects Trypsin 3/PRSS3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Trypsin 3/PRSS3
Applications Western Blot
Classification Monoclonal
Clone 419910
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_035775
Gene Alias Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsin, Mesotrypsinogen, MTG, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS3, PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin 4, Trypsin III, Trypsin IV, trypsin-3, Trypsinogen 15, trypsinogen 4, trypsinogen 5, trypsinogen IV
Gene Symbols PRSS3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Trypsin 3/PRSS3 Phe16-Asn246 Accession # NP_035775
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5646
Test Specificity Detects mouse Trypsin 3/PRSS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, weak cross-reactivity with recombinant human (rh) Trypsin 3 is observed and no cross-reactivity with rhTrypsin 1 or rhTrypsin 2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.