Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Tryptase epsilon/BSSP-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
| Antigen | Tryptase epsilon/BSSP-4/PRSS22 |
|---|---|
| Clone | 246702 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB20592SP
![]() |
R&D Systems
MAB20592SP |
25 μg |
Each for $200.50
|
|
|||||
MAB20592
![]() |
R&D Systems
MAB20592 |
500 μg |
Each for $643.50
|
|
|||||
Description
Tryptase epsilon/BSSP-4/PRSS22 Monoclonal specifically detects Tryptase epsilon/BSSP-4/PRSS22 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Tryptase epsilon/BSSP-4/PRSS22 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSP-2, BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, PRSS22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343 | |
| PRSS22 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 246702 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9ER10 | |
| 64063 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tryptase epsilon/BSSP-4 Ala33-Ser306 Accession # Q9ER10 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Tryptase epsilon/BSSP-4 in ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title