Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TSG-6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2326
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2326 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2326 Supplier R&D Systems Supplier No. BAF2326
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TSG-6 Polyclonal specifically detects TSG-6 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen TSG-6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O08859
Gene Alias Hyaluronate-binding protein, TNF alpha-induced protein 6, TNFAIP6, TNF-stimulated gene 6 protein, TSG6, TSG-6tumor necrosis factor alpha-inducible protein 6, TSG6tumor necrosis factor-inducible gene 6 protein, Tumor necrosis factor alpha-induced protein 6, tumor necrosis factor, alpha-induced protein 6, tumor necrosis factor-stimulated gene-6 protein
Gene Symbols TNFAIP6
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TSG-6 Trp18-Leu275 Accession # O08859
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7130
Test Specificity Detects mouse TSG-6 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human TSG-6 is observed and less than 1% cross-reactivity with recombinant mouse TSG-14 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.