Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSG Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF756SP
This item is not returnable.
View return policy
Description
TSG Polyclonal specifically detects TSG in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TSG | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TSGtwisted gastrulation protein homolog 1, twisted gastrulation homolog 1 (Drosophila), TWSG1 | |
Goat | |
Affinity Purified | |
RUO | |
57045 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9EP52 | |
TWSG1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSG Cys25-Phe222 Accession # Q9EP52 | |
25 μg | |
Primary | |
Detects mouse TSG in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction