Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSLP Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 16 publications
$214.00 - $2110.00
Specifications
Antigen | TSLP |
---|---|
Clone | 152614 |
Dilution | Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Description
TSLP Monoclonal specifically detects TSLP in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
TSLP | |
Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
Q9JIE6 | |
85480 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSLP Tyr20-Glu140 Accession # Q9JIE6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TSLP in Western blots. |
152614 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
thymic stromal lymphopoietin | |
TSLP | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title