Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSLP Antibody, R&D Systems™

Rat Monoclonal Antibody
$227.50 - $549.50
Specifications
Antigen | TSLP |
---|---|
Clone | 152614R |
Dilution | Neutralization 0.1-0.4 ug/mL |
Applications | Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB555RSP
![]() |
R&D Systems
MAB555RSP |
25 μg |
Each for $227.50
|
|
|||||
MAB555R100
![]() |
R&D Systems
MAB555R100 |
100 μg |
Each for $549.50
|
|
|||||
Description
TSLP Monoclonal specifically detects TSLP in Mouse samples. It is validated for Neutralization.Specifications
TSLP | |
Neutralization 0.1-0.4 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
thymic stromal lymphopoietin | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Detects mouse TSLP in direct ELISAs. |
152614R | |
Neutralization | |
Unconjugated | |
RUO | |
Q9JIE6 | |
85480 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title