Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSLPR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF546
This item is not returnable.
View return policy
Description
TSLPR/CRLF2 Polyclonal specifically detects TSLPR/CRLF2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.Specifications
TSLPR/CRLF2 | |
Polyclonal | |
Unconjugated | |
BAA92159 | |
CRLF2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TSLP R Ala20-Leu233 Accession # BAA92159 | |
100 μg | |
Primary | |
Detects mouse TSLP R in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.2-0.6 ug/mL, CyTOF-ready | |
CRL2, CRL2cytokine receptor CRL2 precusor, CRLF2, CRLF2Y, CRLM-2, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, PCOR1, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP R, TSLP receptor, TSLPR | |
Goat | |
Affinity Purified | |
RUO | |
64109 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction