Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSLPR Fluorescein-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB5461F
This item is not returnable.
View return policy
Description
TSLPR/CRLF2 Polyclonal specifically detects TSLPR/CRLF2 in Mouse samples. It is validated for Flow Cytometry.Specifications
TSLPR/CRLF2 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CRL2, CRL2cytokine receptor CRL2 precusor, CRLF2, CRLF2Y, CRLM-2, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, PCOR1, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP R, TSLP receptor, TSLPR | |
Goat | |
Affinity Purified | |
RUO | |
64109 | |
Mouse | |
IgG |
Flow Cytometry | |
Fluorescein | |
BAA92159 | |
CRLF2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TSLP R Ala20-Leu233 Accession # BAA92159 | |
100 Tests | |
Primary | |
Detects mouse TSLP R in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction