Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSPAN8 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB6524S100UG
This item is not returnable.
View return policy
Description
TSPAN8/TM4SF3 Monoclonal specifically detects TSPAN8/TM4SF3 in Mouse samples. It is validated for Flow Cytometry.Specifications
TSPAN8/TM4SF3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CO-029, tetraspanin 8, TM4SF3, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029 | |
Rat | |
100 μg | |
Primary | |
Detects mouse TSPAN8 transfectants but not the parental cell line in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
657909 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
TSPAN8 | |
CHO Chinese hamster ovary cell line transfected with mouse TSPAN8 | |
RUO | |
7103 | |
Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction