Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TSPAN8 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB6524P
This item is not returnable.
View return policy
Description
TSPAN8/TM4SF3 Monoclonal specifically detects TSPAN8/TM4SF3 in Mouse samples. It is validated for Flow Cytometry.Specifications
TSPAN8/TM4SF3 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CO-029, tetraspanin 8, TM4SF3, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7103 | |
Mouse | |
Purified |
Flow Cytometry | |
657909 | |
Flow Cytometry 10 uL/10^6 cells | |
Q8R3G9 | |
TSPAN8 | |
CHO Chinese hamster ovary cell line transfected with mouse TSPAN8 Accession # Q8R3G9 | |
100 Tests | |
Primary | |
Detects mouse TSPAN8 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction