Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Tsukushi/TSK Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6376
Change view
Click to view available options
Quantity:
100 μg
25 μg
Catalog No. Quantity
MAB6376 100 μg
MAB6376SP 25 μg
2 options
This item is not returnable. View return policy

Catalog No. MAB6376

Supplier: R&D Systems MAB6376

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Tsukushi/TSK Monoclonal specifically detects Tsukushi/TSK in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Tsukushi/TSK
Applications Western Blot
Classification Monoclonal
Clone 462310
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8CBR6
Gene Alias E2IG4, E2IG4E2-induced gene 4 protein, leucine rich repeat containing 54, Leucine-rich repeat-containing protein 54, LRRC54, TSKtsukushi homolog, TSKU, Tsukushi, tsukushi small leucine rich proteoglycan homolog (Xenopus laevis), tsukushi, small leucine rich proteoglycan, tsukushin
Gene Symbols TSKU
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Tsukushi/TSK Thr18-Leu354 Accession # Q8CBR6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25987
Test Specificity Detects mouse Tsukushi/TSK in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Tsukushi/TSK is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2c
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.