Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TWEAK R/TNFRSF12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1610
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1610 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1610 Supplier R&D Systems Supplier No. AF1610
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TWEAK R/TNFRSF12 Polyclonal specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TWEAK R/TNFRSF12
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. Q9CR75
Gene Alias CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14
Gene Symbols TNFRSF12A
Host Species Goat
Immunogen E. coli-derived recombinant mouse TWEAK R/TNFRSF12 Glu28-Trp79 Accession # Q9CR75
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51330
Test Specificity Detects mouse TWEAK R in ELISAs and Western blots. In sandwich ELISAs, less than 2% cross-reactivity with recombinant human TWEAK R is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.