Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Twist-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB183434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183434 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183434 Supplier Novus Biologicals™ Supplier No. NBP306810
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Cell Culture Supernatant, Plasma, Serum
Specificity This kit recognizes Mouse Twist-1 in samples. No significant cross-reactivity or interference between Mouse Twist-1 and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 7291
Gene Symbol TWIST1
Quantity 1 Kit
Research Discipline Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, twist-related protein 1
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.