Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Tyro3/Dtk Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB759
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| Dtk/TYRO3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7301 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, CyTOF, ELISA | |
| 109646 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P55144 | |
| Tyro3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk Ala31-Ser418 Accession # P55144 | |
| 500 μg | |
| Primary | |
| Detects mouse Tyro3/Dtk extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Tyro3/Dtk, recombinant mouse (rm) Mer, or rmAxl is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction