Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Tyro3/Dtk Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF759
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF759 Supplier R&D Systems Supplier No. AF759
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 8 publications

Dtk/TYRO3 Polyclonal specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Dtk/TYRO3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P55144
Gene Alias Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Gene Symbols Tyro3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk Ala31-Ser418 Accession # P55144
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7301
Test Specificity Detects mouse Tyro3/Dtk in direct ELISAs and Western blots. In direct ELISA and Western blots, approximately 20% cross-reactivity with recombinant human (rh) Tyro3/Dtk is observed, and less than 1% cross-reactivity with recombinant mouse (rm) Axl, rmMer and rhAxl is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.