Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse u-Plasminogen Activator (uPA)/Urokinase Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9185100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9185100 100 μg
MAB9185SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9185100 Supplier R&D Systems Supplier No. MAB9185100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen u-Plasminogen Activator/Urokinase
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 901420
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P06869
Gene Alias ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse uPA Met1-Phe433 Accession # P06869
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5328
Test Specificity Detects mouse uPA in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.