Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse UGRP1/SCGB3A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3465
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3465 100 μg
MAB3465SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3465 Supplier R&D Systems Supplier No. MAB3465
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

UGRP1/SCGB3A2 Monoclonal specifically detects UGRP1/SCGB3A2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen UGRP1/SCGB3A2
Applications Western Blot
Classification Monoclonal
Clone 381707
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q920H1
Gene Alias LU103, Pneumo secretory protein 1, PnSP-1, PNSP1secretoglobin family 3A member 2, SCGB3A2, secretoglobin, family 3A, member 2, UGRP1pnSP-1, Uteroglobin-related protein 1
Gene Symbols SCGB3A2
Host Species Rat
Immunogen E. coli-derived recombinant mouse UGRP1/SCGB3A2 isoform C Leu22-Leu139 Accession # Q920H1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117156
Test Specificity Detects mouse UGRP1/SCGB3A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human UGRP1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.