Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse UGRP1/SCGB3A2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3465SP
This item is not returnable.
View return policy
Description
UGRP1/SCGB3A2 Polyclonal specifically detects UGRP1/SCGB3A2 in Mouse samples. It is validated for Western Blot.Specifications
UGRP1/SCGB3A2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LU103, Pneumo secretory protein 1, PnSP-1, PNSP1secretoglobin family 3A member 2, SCGB3A2, secretoglobin, family 3A, member 2, UGRP1pnSP-1, Uteroglobin-related protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
117156 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q920H1 | |
SCGB3A2 | |
E. coli-derived recombinant mouse UGRP1/SCGB3A2 isoform C Leu22-Leu139 Accession # Q920H1 | |
25 μg | |
Primary | |
Detects mouse UGRP1/SCGB3A2 isoforms C and A in direct ELISAs and Western blots. It is also expected to recognize isoform B in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human UGRP1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction