Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ULBP-1/MULT-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2588V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2588V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2588V100 Supplier R&D Systems Supplier No. FAB2588V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

ULBP-1 Monoclonal specifically detects ULBP-1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ULBP-1
Applications Flow Cytometry
Classification Monoclonal
Clone 237104
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1
Gene Symbols ULBP1
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with mouse ULBP-1MULT-1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80329
Test Specificity Detects mouse ULBP-1MULT-1.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.