Learn More
Novus Biologicals™ Mouse Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Universal |
| Quantity | 4 Vials |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Universal Protein Lysate: Mouse Normal Tissues |
| Cellular Fraction | Whole |
| Tissue Condition | Normal |
| Cross Reactivity | Mouse |
| Host Species | Mouse |
| Concentration | 5 mg/mL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.