Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse uPAR Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$200.50 - $2070.00
Specifications
Antigen | uPAR |
---|---|
Clone | 109801 |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-reported |
Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Classification | Monoclonal |
Description
uPAR Monoclonal specifically detects uPAR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-reported.Specifications
uPAR | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-reported | |
Monoclonal | |
Rat | |
Mouse | |
P35456 | |
5329 | |
Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1 Leu24-Thr297 Accession # P35456 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse uPAR in Western blots. In Western blots, no cross-reactivity with recombinant human uPAR is observed. |
109801 | |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, PLAUR, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
PLAUR | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title