Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Urotensin-II R Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB9245T100UG
This item is not returnable.
View return policy
Description
Urotensin-II R/GPR14 Monoclonal specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry.Specifications
Urotensin-II R/GPR14 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Rat | |
100 μg | |
Primary | |
Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
443707 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R | |
Mouse Urotensin-II R synthetic peptide | |
RUO | |
2837 | |
Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction