Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Urotensin-II R Alexa Fluor 647-conjugated Antibody, R&D Systems™

Catalog No. FAB9245R025
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9245R025 25 μg
FAB9245R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB9245R025 Supplier R&D Systems Supplier No. FAB9245R025UG
This item is not returnable. View return policy

Rat Monoclonal Antibody

Urotensin-II R/GPR14 Monoclonal specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Urotensin-II R/GPR14
Applications Flow Cytometry
Classification Monoclonal
Clone 443707
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.2 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R
Host Species Rat
Immunogen Mouse Urotensin-II R synthetic peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2837
Test Specificity Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.