Learn More
Mouse Urotensin-II R Alexa Fluor 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Urotensin-II R/GPR14 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 443707 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.2 ug/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R |
| Host Species | Rat |
| Immunogen | Mouse Urotensin-II R synthetic peptide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.