Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Urotensin-II R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7121429
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9245SP 25 μg
MAB9245100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9245SP Supplier R&D Systems Supplier No. MAB9245SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Urotensin-II R/GPR14 Monoclonal specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Urotensin-II R/GPR14
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 443707
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R
Host Species Rat
Immunogen Mouse Urotensin-II R synthetic peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2837
Test Specificity Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.