Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VCAM-1/CD106 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6432100
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
VCAM-1/CD106 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7412 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF, ELISA | |
112734 | |
Western Blot 2 ug/mL, Simple Western 100 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P29533 | |
VCAM1 | |
Mouse myeloma cell line NS0-derived recombinant mouse VCAM-1 Phe25-Glu698 (predicted) Accession # P29533 | |
100 μg | |
Primary | |
Detects recombinant mouse (rm) VCAM-1 in Western blots and ELISAs. In sandwich immunoassays, no cross-reactivity or interference was observed with recombinant human VCAM-1, rmICAM-1, rmE-Selectin, rmL-Selectin or rmP-Selectin. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction