Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VE-Cadherin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1002
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1002 Supplier R&D Systems Supplier No. MAB1002
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

VE-Cadherin Monoclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen VE-Cadherin
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 162709
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. 2208309A
Gene Alias 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin
Gene Symbols CDH5
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin Asp46-Gln592 (Gly67 del, Ile69Asp, Lys70Gln) Accession # 2208309A
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1003
Test Specificity Detects mouse VE-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-17, rhN-Cadherin, recombinant mouse (rm) E-Cadherin, or rmP-Cadherin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.