Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VE-Cadherin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1002
This item is not returnable.
View return policy
Description
VE-Cadherin Polyclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot.Specifications
VE-Cadherin | |
Polyclonal | |
Biotin | |
2208309A | |
CDH5 | |
Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin Asp46-Gln592 Accession # 2208309A | |
50 μg | |
Primary | |
Detects mouse VE-Cadherin in Western blots. In this format, less than 5% cross-reactivity with recombinant human VE-Cadherin is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
Goat | |
Affinity Purified | |
RUO | |
1003 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction