Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VEGF-B186 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF767
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF767 100 μg
AF767SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF767 Supplier R&D Systems Supplier No. AF767
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

VEGF-B Polyclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen VEGF-B
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Alias vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Gene Symbols Vegfa
Host Species Goat
Immunogen E. coli-derived recombinant mouse VEGF-B 186
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7423
Test Specificity Detects mouse VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) VEGF-B 186 is observed and no cross-reactivity with rhVEGF 165 recombinant mouse (rm) VEGF-B 167 rhVEGF-C, and rmVEGF-D is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.