Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VEGF-D Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF469
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF469 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF469 Supplier R&D Systems Supplier No. BAF469
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

VEGF-D Polyclonal specifically detects VEGF-D in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen VEGF-D
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P97946
Gene Alias c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor
Gene Symbols FIGF
Host Species Goat
Immunogen E. coli-derived recombinant mouse VEGF-D Phe98-Ser206 Accession # P97946
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2277
Test Specificity Detects mouse VEGF-D in ELISAs and Western blots. In sandwich immunoassays, less than 75% cross-reactivity with recombinant human (rh) VEGF-D is observed and less than 0.1% cross-reactivity with recombinant mouse (rm) VEGF, rmVEGF-B, and rhVEGF-C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.