Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VEGF120/164 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB493SP
This item is not returnable.
View return policy
Description
VEGF Monoclonal specifically detects VEGF in Mouse samples. It is validated for Western Blot.Specifications
VEGF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7422 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
39917 | |
Western Blot 1 ug/mL | |
AAA40547 | |
VEGFA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGF 164 Ala27-Arg190 Accession # AAA40547 | |
25 μg | |
Primary | |
Detects mouse VEGF 120/164 in direct ELISAs and Western blots. This antibody does not recognize recombinant mouse VEGF 115 or any isoform of human VEGF. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction