Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Vegfc (P97953, 125 amino acids) Partial Recombinant Protein

Catalog No. 89967016 Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg

Used for Func, SDS-PAGE

Specifications

Accession Number P97953
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 22341
Molecular Weight (g/mol) 15kDa
Name Vegfc (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 0.1% acetic acid, store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias AW228853/VEGF-C
Common Name Vegfc
Gene Symbol Vegfc
Biological Activity Activity of VEGF-C will be tested by the stimulation of VEGF-R3 phosphorylation in cells transfected with VEGF-R3. The expected ED50 is > 200ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.