Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse VEGFR1/Flt-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB4711SP
This item is not returnable.
View return policy
Description
VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
VEGFR1/Flt-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2321 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Flow Cytometry, CyTOF | |
141522 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P35969 | |
FLT1 | |
Mouse myeloma cell line NS0-derived recombinant mouse VEGFR1/Flt-1, Ser27-Glu759, Accession # P35969 | |
25 μg | |
Primary | |
Detects mouse VEGFR1/Flt-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human VEGFR1, recombinant mouse (rm) VEGFR2, or rmVEGFR3 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction