Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VEGFR3/Flt-4 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF743 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF743 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF743 Supplier R&D Systems Supplier No. AF743
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 58 publications

VEGFR3/Flt-4 Polyclonal specifically detects VEGFR3/Flt-4 in Cynomolgus Monkey∣Equine∣Human∣Mouse∣Rat∣Transgenic Mouse samples. It is validated for CyTOF∣ELISA Development∣Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣Western Blot.

Specifications

Antigen VEGFR3/Flt-4
Applications CyTOF, ELISA Development, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. P35917
Gene Alias EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Gene Symbols FLT4
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGFR3/Flt-4, Tyr25-Asp770, Accession # P35917
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2324
Test Specificity Detects mouse VEGFR3/Flt-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) VEGFR3 is observed and less than 5% cross-reactivity with recombinant mouse VEGFR2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Cynomolgus Monkey, Equine, Human, Mouse, Rat, Transgenic Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.