Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse VEGFR3/Flt-4 ELISA Kit (Colorimetric)
SDP

Catalog No. NB009569
Click to view available options
Quantity:
1 Kit

This kit recognizes Mouse VEGFR3/Flt4 in samples.

No Significant cross-reactivity or interference between Mouse VEGFR3/Flt4 and analogues was observed.

Specifications

Accession Number P35917, P35917
Assay Range 62.50 - 4000 pg/mL
Assay Sensitivity 37.50 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 2324
Gene Symbol FLT4
Host Species Mouse
Quantity 1 Kit
Research Discipline Angiogenesis, Cancer, Hypoxia, Protein Kinase
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target VEGFR3/Flt-4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.