Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse VIP ELISA Kit (Colorimetric)
SDP

Catalog No. NB030368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB030368 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB030368 Supplier Novus Biologicals™ Supplier No. NBP282439
Add to Cart
Add to Cart

Mouse VIP ELISA Kit (Colorimetric) measures Mouse VIP in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 12.5 to 800 pg/mL
Assay Sensitivity 7.5 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse VIP in samples. No Significant cross-reactivity or interference between Mouse VIP and analogues was observed.
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 7432
Gene Symbol VIP
Quantity 1 Kit
Research Discipline Cancer, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides
Target VIP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.