Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Vitamin D BP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB41881100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB41881100 100 μg
MAB41881SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB41881100 Supplier R&D Systems Supplier No. MAB41881100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Vitamin D BP Monoclonal specifically detects Vitamin D BP in Mouse samples. It is validated for ELISA.

Specifications

Antigen Vitamin D BP
Applications ELISA
Classification Monoclonal
Clone 884706
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P21614
Gene Alias DBP, DBP/GC, DBP-MAF, DBPVDBG, EC 6.3.1.5, Gc, Gc-globulin, Gc-MAF, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBG, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Vitamin D BP, Met1-Ser476, Accession # P21614
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2638
Test Specificity Detects mouse Vitamin D BP in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.