Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Vitamin D BP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4188
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4188 Supplier R&D Systems Supplier No. AF4188
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Vitamin D BP Polyclonal specifically detects Vitamin D BP in Mouse samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Vitamin D BP
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Gene Accession No. P21614
Gene Alias DBP, DBP/GC, DBP-MAF, DBPVDBG, EC 6.3.1.5, Gc, Gc-globulin, Gc-MAF, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBG, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Gene Symbols GC
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Vitamin D BP Leu17-Ser476 Accession # P21614
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2638
Test Specificity Detects mouse Vitamin D BP in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human Vitamin D BP is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.