Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Vitronectin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
Supplier: R&D Systems MAB38751SP
This item is not returnable.
View return policy
Description
Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for Immunohistochemistry.Specifications
Vitronectin | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
AAH12690 | |
VTN | |
Mouse myeloma cell line NS0-derived recombinant mouse Vitronectin Asp20-Lys478 Accession # AAH12690 | |
25 μg | |
Primary | |
Detects mouse Vitronectin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Vitronectin is observed. | |
Mouse | |
Purified |
Immunohistochemistry | |
347317 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT, VTN | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7448 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction