Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VSIG2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4766SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4766SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4766SP Supplier R&D Systems Supplier No. MAB4766SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

VSIG2 Monoclonal specifically detects VSIG2 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen VSIG2
Applications Immunohistochemistry
Classification Monoclonal
Clone 771625
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z109
Gene Alias 2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Gene Symbols VSIG2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse VSIG2 Val25-Ala244 (Glu240Asp) Accession # Q9Z109
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23584
Test Specificity Detects mouse VSIG2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human VSIG2 or recombinant mouse VSIG1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.