Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VSIG2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4766
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4766 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4766 Supplier R&D Systems Supplier No. BAF4766
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

VSIG2 Polyclonal specifically detects VSIG2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen VSIG2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9Z109
Gene Alias 2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Gene Symbols VSIG2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse VSIG2 isoform 1 Val25-Ala244 Accession # Q9Z109
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23584
Test Specificity Detects mouse VSIG2 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) VSIG2 is observed and less than 10% cross-reactivity with rhVSIG3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.