Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse WARP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4927
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4927 100 μg
AF4927SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4927 Supplier R&D Systems Supplier No. AF4927
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

WARP/VWA1 Polyclonal specifically detects WARP/VWA1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen WARP/VWA1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q8R2Z5
Gene Alias DKFZp761O051, von Willebrand factor A domain containing 1, von Willebrand factor A domain-containing protein 1, von Willebrand factor A domain-related protein, VWA1, WARP
Gene Symbols VWA1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse WARP isoform 1 Arg19-Pro415 Accession # Q8R2Z5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64856
Test Specificity Detects mouse WARP in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human WARP is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.