Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse WIF-1 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

$200.50 - $2350.00

Specifications

Antigen WIF-1
Clone 134714
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB135SP
GREENER_CHOICE
View Documents
R&D Systems
MAB135SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB135050
GREENER_CHOICE
View Documents
R&D Systems
MAB135050
50 μg
Each for $292.00
Only null left
Add to Cart
 
MAB135500
GREENER_CHOICE
View Documents
R&D Systems
MAB135500
500 μg
Each for $2,350.00
Only null left
Add to Cart
 
Description

Description

WIF-1 Monoclonal specifically detects WIF-1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

WIF-1
LYOPH
Western Blot
Unconjugated
Rat
Mouse
Q9WUA1
11197
Mouse myeloma cell line NS0-derived recombinant mouse WIF-1 Gly29-Trp379 Accession # Q9WUA1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse WIF-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human WIF-1 is observed.
134714
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
WIF1, WIF-1, WNT inhibitory factor 1
WIF1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.