Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse WISP-1/CCN4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1680100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1680100 100 μg
MAB1680SP 25 μg
MAB1680500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1680100 Supplier R&D Systems Supplier No. MAB1680100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

WISP-1/CCN4 Monoclonal specifically detects WISP-1/CCN4 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen WISP-1/CCN4
Applications Western Blot
Classification Monoclonal
Clone 214203
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O54775
Gene Alias CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1
Gene Symbols WISP1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse WISP-1/CCN4 Leu21-Asn367 Accession # O54775
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8840
Test Specificity Detects mouse WISP-1/CCN4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 10-30% cross-reactivity with recombinant human (rh) WISP-1 but no cross-reactivity with rhCTGF or rmNOV.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.