Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1620SP
This item is not returnable.
View return policy
Description
Wnt-1 Polyclonal specifically detects Wnt-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Int-1, INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1), Wnt1 | |
Goat | |
Affinity Purified | |
RUO | |
7471 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q3UR96 | |
WNT1 | |
E. coli-derived recombinant mouse Wnt-1 peptide Ser52-Glu92 and Met234-Arg313 Accession # Q3UR96 | |
25 μg | |
Primary | |
Detects mouse Wnt-1 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-3a and rmWnt-5a is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction