Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-11 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3746SP
This item is not returnable.
View return policy
Description
Wnt-11 Monoclonal specifically detects Wnt-11 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-11 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HWNT11, MGC141946, MGC141948, protein Wnt-11, wingless-type MMTV integration site family, member 11, Wnt11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7481 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunohistochemistry | |
310622 | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
NP_033545 | |
WNT11 | |
E. coli-derived recombinant mouse Wnt-11 Leu39-Ala79 and Ser225-Arg297 Accession # NP_033545 | |
25 μg | |
Primary | |
Detects mouse Wnt-11 in direct ELISAs and Western blots. In these formats, this antibody shows no cross-reactivity with recombinant human Wnt-2, -7a, -7b, recombinant mouse Wnt-1, -3a, -4, -5a, -5b, -8a, -8b, or û9a. Western blotting with human cell extracts demonstrates that this antibody also detects human Wnt-11. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction