Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Wnt-3a DuoSet ELISA

Catalog No. dy1324b05
Click to view available options
Quantity:
1 Kit (5 Plates)
This item is not returnable. View return policy
This item is not returnable. View return policy

Mouse Wnt-3a DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Wnt-3a DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies Wnt-3a.

Specifications

Assay Range 156.00 to 10000 pg/mL
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 89780
Host Species Mouse
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.