Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF475
This item is not returnable.
View return policy
Description
Wnt-4 Polyclonal specifically detects Wnt-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-4 | |
Polyclonal | |
Unconjugated | |
P22724 | |
WNT4 | |
E. coli-derived recombinant mouse Wnt-4 Ile37-Glu76, Arg222-Thr295 Accession # P22724 | |
100 μg | |
Primary | |
Detects mouse Wnt-4 in direct ELISAs and Western blots. The antibody has also been shown to recognize a mammalian cell-expressed mouse Wnt-4 protein in Western blots. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
protein Wnt-4, SERKAL, wingless-type MMTV integration site family, member 4, Wnt4, WNT-4 | |
Goat | |
Affinity Purified | |
RUO | |
54361 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction