Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-5b Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3006SP
This item is not returnable.
View return policy
Description
Wnt-5b Monoclonal specifically detects Wnt-5b in Mouse samples. It is validated for Immunocytochemistry.Specifications
Wnt-5b | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
NP_033551 | |
WNT5B | |
E. coli-derived recombinant mouse Wnt-5b Ser55-Ser95 and Lys246-Glu315 connected by a Gly-Ser linker Accession # NP_033551 | |
25 μg | |
Primary | |
Detects mouse Wnt-5b in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Wnt-5a, -8a, -8b, -9b, -10a, -10b, or recombinant human Wnt-7b is observed. | |
Mouse | |
Purified |
Immunocytochemistry | |
325004 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, Wnt5b, WNT-5B protein | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
81029 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction